Learn More
Sigma-Transwab Liquid Amies Media Swabs with Swab Capture
Liquid transport for automated processing systems and conventional inoculation and plating
Manufacturer:  ADVANTAGE BUNDLING SP LLC MW176S
 View more versions of this product
Catalog No. 23-691-017
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Medical Wire Swab
Polyurethane
Screw cap
For aerobic, anaerobic and fastidious microorganisms
5.9 in.
Conical
CE, M-40
Purple
Liquid amies medium
15cm
Safety and Handling
ShelfLife : 2 Year(s)