Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Sign: CONFINED SPACE

Improve workplace safety with appropriate signage. The Brady™ Confined Space Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

Manufacturer:  Brady™ 60569

 View more versions of this product

Catalog No. 19-107-875


  Check Availability
Add to cart

Specifications

Description

  • Easily mounted solutions in a variety of environments
  • Brady™ signs feature bright colors, bold text, and intuitive pictographs
  • Highly visible and easily understood
Description & Specifications

Specifications

Black/red on white
10 in.
CONFINED SPACE
Rectangle
-40°F to 230°F
14 in.
Laminated
Polyester with polyester overlaminate
1/Sign
14 in. x 10 in. x 0.01 in.
0.01 in.
Self-adhesive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit