Learn More
Brady™ Sign: CONFINED SPACE ENTER BY PERMIT ONLY...OBTAIN AUTHORIZED PERMIT
Improve workplace safety with signage. The Brady™ Confined Space Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$24.70 - $25.50
Specifications
Dimensions (L x W) | 10 x 7 in. |
---|---|
Shape | Rectangular |
Displayed Text | CONFINED SPACE ENTER BY PERMIT ONLY PREPARE FOR ENTER BY PERMIT ONLY PREPARE FOR ENTRY IDENTIFY HAZARDS OF PERMIT SPACE. DE-ENERGIZE AND LOCKOUT ALL ENERGY SOURCES. DRAIN, CLEAN AND VENTILATE CONFINED SPACES. ISOLATE CONFINED-DISCONNECT FILL AND DRAIN LINES TEST ATMOSPHERE OXYGEN LEVEL BETWEEN 19.5% AND 23.5%. FLAMMABLE GLASS/VAPORS LESS THAN 10% ID LEL. ALL SUBSTANCES BELOW ESTABLISHED PEL. PREPSARE PERSONAL PROTECTION DEVICES RESPIRATOR, PROTECTIVE CLOTHING, LIFELINE AND SAFETY HAMESS. ATTENDANT AND RESCUE EQUIPENT IN PLACE REVIEW COMMUNICATION REOCEDURES OBTAIN AUTHORIZED PERMIT |
Temperature Range (English) | -40°F to 230°F |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Mounting Type | Dimensions (L x W) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mounting Type | Dimensions (L x W) | Material | Price | Quantity & Availability | |||
19-107-787
|
Brady™
122405 |
Corner holes | 10 x 7 in. | Aluminum |
Each for $24.70
|
|
|||
19-107-788
|
Brady™
122649 |
Self-adhesive | 10 x 7 in. | Polyester |
Each for $25.50
|
|
|||
Description
- Abrasion-resistant and chemical-resistant
- Brady™ signs feature bright colors, bold text, and intuitive pictographs
- Highly visible and easily understood
Specifications
10 x 7 in. | |
CONFINED SPACE ENTER BY PERMIT ONLY PREPARE FOR ENTER BY PERMIT ONLY PREPARE FOR ENTRY IDENTIFY HAZARDS OF PERMIT SPACE. DE-ENERGIZE AND LOCKOUT ALL ENERGY SOURCES. DRAIN, CLEAN AND VENTILATE CONFINED SPACES. ISOLATE CONFINED-DISCONNECT FILL AND DRAIN LINES TEST ATMOSPHERE OXYGEN LEVEL BETWEEN 19.5% AND 23.5%. FLAMMABLE GLASS/VAPORS LESS THAN 10% ID LEL. ALL SUBSTANCES BELOW ESTABLISHED PEL. PREPSARE PERSONAL PROTECTION DEVICES RESPIRATOR, PROTECTIVE CLOTHING, LIFELINE AND SAFETY HAMESS. ATTENDANT AND RESCUE EQUIPENT IN PLACE REVIEW COMMUNICATION REOCEDURES OBTAIN AUTHORIZED PERMIT | |
1 Ea. | |
230°F | |
10 in. |
Rectangular | |
-40°F to 230°F | |
7 in. | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.