Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Bluing Reagent

Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.

$146.40 - $451.50

Specifications

Product Type Bluing Reagent
Format Liquid
pH 8
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-050-114
Accessories View Documents Promotion Details
Epredia™
7301
1 gal.
Each for $148.40
Edge
Add to cart
 
73-01L
View Documents Promotion Details
Epredia™
7301L
1.32 gal.
Each for $146.40
Add to cart
 
22-050-115
Accessories View Documents Promotion Details
Epredia™
7341
4 x 1 gal.
Case of 4 for $451.50
Add to cart
 
Description

Description

  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
  • Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
Specifications

Specifications

Bluing Reagent
Liquid
8
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit