Learn More
Epredia™ Signature Series Cover Glass
Eliminate sticking and reduce waste with the uniquely manufactured Epredia™ Signature Series Cover Glass. Ideal for automatic coverslippers.
Supplier: Epredia™ 12450S
This item is not returnable.
View return policy
Description
Thickness 1:- 0.13 - 0.17mm
- Case of 10 Pk. (68-72/pack)
- Available in several sizes
Specifications
24 mm | |
#1 (0.13 to 0.16 mm) | |
Case of 10 Pk. (68-72/pack) |
50 mm | |
50 x 24 mm |