Learn More
Thermo Scientific™ Signature Series Cover Glasses Available on GSA/VA Contract for Federal Government customers only.
Eliminate sticking and reduce waste with the uniquely manufactured Thermo Scientific™ Signature Series Cover Glass. Ideal for automatic coverslippers.
$295.50 - $300.50
Specifications
Length (Metric) 24mm
Thickness (Metric) 0.13 to 0.17mm
View More Specs
Products 3
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
22-050-231
View Documents
Thermo Scientific™
12440S
40mm Case of 10 for $300.50
Add to cart
22-050-232 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
12450S
50mm Case of 10 for $295.50
Add to cart
22-050-233 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
12460S
60mm Case of 10 for $295.50
Add to cart
Description
Description
Eliminate sticking and reduce waste with the uniquely manufactured Thermo Scientific™ Signature Series Cover Glass. Ideal for automatic coverslippers.
Thickness 1:
  • 0.13 - 0.17mm
Ordering Information:
  • Case of 10 Pk. (68-72/pack)
  • Available in several sizes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
24mm
0.13 to 0.17mm