Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Signature Series™ Hematoxylin 2
Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.
$684.00 - $956.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-050-113
|
Epredia™
7231 |
1 pt. |
Case of 4 for $684.00
|
|
|||||
72-31L
|
Epredia™
7231L |
1.32 gal. |
Each for $956.00
|
|
|||||
Description
- No filtration required
- Stains up to 2500 slides per pint
- Part of the Signature Series Staining System
- Crisp, well-defined nuclear detail chromatin delineation
Available in 5L packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Hematoxylin 2 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title