Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK2 Mouse anti-Human, Clone: S15G10, Abnova™
Mouse monoclonal antibody raised against recombinant human SIK2.
Supplier: Abnova Corporation MAB12947
Specifications
SIK2 | |
Monoclonal | |
Unconjugated | |
Immunocytochemistry Immunofluorescence (1:100-1:400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1:250) The optimal working dilution should be determined by the end user. | |
DKFZp434K1115/KIAA0781/LOH11CR1I/QIK/SNF1LK2 | |
Mouse | |
Affinity purification | |
RUO | |
23235 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
S15G10 | |
Mouse monoclonal antibody raised against recombinant human SIK2. | |
In PBS, pH 7.2 (0.09% sodium azide). | |
SIK2 | |
Recombinant protein corresponding to human SIK2. | |
50 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction