Learn More
SIL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 241101AP
Catalog No. 50-173-4239
Add to cart
Description
Description
This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.
Specifications
Specifications
SIL1
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPK6, Q9H173
BAP, BiP associated protein, MSS, SIL1, ULG5
Rabbit
IgG
150 μL
-20°C
Primary
64374, 81500
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SIL1
Liquid
SIL1
SIL1 Fusion Protein Ag21367
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse