Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silaris™ Dock for use with Silaris in-vitro diagnostic assays

Click to view available options
For Use With (Application):
Silaris In-Vitro Diagnostic Assays

The Silaris Dock is a reusable platform, used in conjunction with the Silaris Test Cassettes, to run in-vitro diagnostic assays.

The Silaris Dock is a simple to use, requires no calibration, and no maintenance or service contracts, that is compact and portable for use wherever testing is needed. 

Temperature Range (Metric) 15°C to 30°C
For Use With (Application) Silaris In-Vitro Diagnostic Assays
Frequency 50/60 Hz
Height (English) 5.7 in.
Height (Metric) 14.5 cm
Includes 1 Silaris Dock, 1 AC Power Adaptor, 1 AC Power Cord, 1 Dock Operators Manual
Length (English) 3.9 in.
Length (Metric) 9.7 cm
Timer Internal Timer
Type Dock
Voltage 12 VDC ± 1 VDC
Weight (Metric) 619 g
Width (English) 3.9 in.
Width (Metric) 9.7 cm
Show More Show Less