Learn More
Sekisui Diagnostics Silaris™ Influenza A&B Control  Kit
The Silaris™ Influenza  A&B Control  Kit is intended for in vitro diagnostic use in quality control testing with the Silaris Influenza A&B Test.
Manufacturer:  Sekisui Diagnostics 1024
Catalog No. 22-321-117
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Specifications
Specifications
No
Influenza A&B
Each
5 Influenza A+/B- Control Swab, 5 Influenza A-/B+ Control Swab, 1-Instructions For Use
10
15° - 30°C
N/A
Silaris Flu A&B Test
N/A
2.25 in. x 5.25 in. x 2.75 in.
2.25 in. x 5.25 in. x 2.75 in.
N/A