Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui Diagnostics Silaris™ Influenza A&B Test Kit, 25/PK

The CLIA Waived Silaris™ Influenza A&B Test performed on the Silaris Dock is a molecular in vitro diagnostic test utilizing (PCR) and lateral flow technology for the qualitative, visual detection and differentiation of influenza A&B.

Manufacturer:  Sekisui Diagnostics 1027

Catalog No. 22-321-116



Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

The CLIA Waived Silaris™ Influenza A&B Test is


ACCURATE

  • Molecular Results
  • Sensitivity of 97% Flu A and 94% Flu B
  • Specificity of 94% Flu A and 99% Flu B
  • Accuracy exceeding conventional rapid lateral flow immunoassays.

AFFORDABLE

  • No capital equipment costs
  • Controls are provided in each kit

SIMPLE

  • Easy to use
  • No calibration or maintenance of the Dock
  • Direct nasal swab sample collection
Specifications

Specifications

No
Influenza A&B
Kit
25 Test Cassetes, 25 Nasal Swab Buffer, 1 Influenza A+/B- Control Swab, 1 Influenza A-/B+ Control Swab, 25 Rayon Swabs 1 Quick Reference Guide, 1 Instructions For Use
25
15° - 30°C
N/A
Silaris Dock
N/A
4 in. x 12 in. x 12 in.
4 in. x 12 in. x 12 in.
N/A
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit