Learn More
Sekisui Diagnostics Silaris™ RSV Test Kit
The CLIA Waived Silaris™ RSV Test is a molecular diagnostic test that utilizes polymerase chain reaction (PCR) technology, providing accurate results to aid in early diagnosis and proper management of RSV.
Manufacturer:  Sekisui Diagnostics 1030
Catalog No. 22-321-119
Add to cart
This item is not returnable. View return policy
Description
Description
  • Accurate
  • Molecular results
  • State-of-the art microfluidic cassette technology
  • PCR test using proprietary OscAR™ Technology
  • No age limitations
  • Affordable
  • Results from a single test
  • Controls in every kit
  • Simple
  • Easy to use
  • Nasal swab collection
  • Compact portable dock with no calibration required
  • Room temperature storage
Specifications
Specifications
Silaris™, Silaris™ RSV Control Kit
25/Pk
Store at room temperature