Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sildenafil Citrate, USP, 98-102%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
171599-83-0
C28H38N6O11S
666.70
DEIYFTQMQPDXOT-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid; 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1
171599-83-0
C28H38N6O11S
666.70
DEIYFTQMQPDXOT-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid; 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

S3092, 171599-83-0, C22H30N6O4S•C6H8O7

Spectrum™ Chemical Sildenafil Citrate, USP
CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 102%
Grade USP