Learn More
Selleck Chemical LLCSupplier Diversity Partner Sildenafil Citrate 50mg 171599-83-0  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S143150MG
Sildenafil Citrate, a selective inhibitor of cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5), is a well-tolerated and highly effective treatment for erectile dysfunction.
Catalog No. 50-631-9