Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ C18 (17% C) Monomeric nec, 40 to 63 μm, 60 Å

SiliaBond™ C18 (17% C) Monomeric nec, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.

$33.10 - $2440.00

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C18 Monomeric nec
Technique Column Chromatography
Packaging Bulk
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-182-237
View Documents
SiliCycle™
R33330B5G
5 g
Each for $33.10
Add to Cart
 
01-182-231
View Documents
SiliCycle™
R33330B10G
10 g
Each for $52.30
Add to Cart
 
01-182-234
View Documents
SiliCycle™
R33330B25G
25 g
Each for $102.90
Add to Cart
 
01-182-236
View Documents
SiliCycle™
R33330B50G
50 g
Each for $191.10
Add to Cart
 
01-182-230
View Documents
SiliCycle™
R33330B100G
100 g
Each for $306.00
Add to Cart
 
01-182-233
View Documents
SiliCycle™
R33330B250G
250 g
Each for $716.50
Add to Cart
 
01-182-235
View Documents
SiliCycle™
R33330B500G
500 g
Each for $1,352.00
Add to Cart
 
01-182-232
View Documents
SiliCycle™
R33330B1KG
1 kg
Each for $2,440.00
Add to Cart
 
Description

Description

  • Purification of low to high polarity compounds.
  • Reproducible purification without complexity and cost of preparative HPLC.
Specifications

Specifications

40 to 63 μm
C18 Monomeric nec
Bulk
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
15.5%
60 Å
Column Chromatography
C18 (Octadecyl, ODS)
SiliaBond™
Silica Gel
Purification of low to medium polarity compounds
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit