Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ C18 (17% C) Polymeric, 40 to 63 μm, 60 Å

SiliaBond™ C18 (17% C) Polymeric, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.

$26.25 - $1979.00

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C18 Polymeric
Technique Column Chromatography
Packaging Bulk
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-182-180
View Documents
SiliCycle™
R00230B5G
5 g
Each for $26.25
Add to Cart
 
01-182-174
View Documents
SiliCycle™
R00230B10G
10 g
Each for $42.55
Add to Cart
 
01-182-177
View Documents
SiliCycle™
R00230B25G
25 g
Each for $84.00
Add to Cart
 
01-182-179
View Documents
SiliCycle™
R00230B50G
50 g
Each for $155.40
Add to Cart
 
01-182-173
View Documents
SiliCycle™
R00230B100G
100 g
Each for $248.00
Add to Cart
 
01-182-176
View Documents
SiliCycle™
R00230B250G
250 g
Each for $601.00
Add to Cart
 
01-182-178
View Documents
SiliCycle™
R00230B500G
500 g
Each for $1,100.00
Add to Cart
 
01-182-175
View Documents
SiliCycle™
R00230B1KG
1 kg
Each for $1,979.00
Add to Cart
 
Description

Description

Purification of low to high polarity compounds. Reproducible purification without complexity and cost of preparative HPLC.

Specifications

Specifications

40 to 63 μm
C18 Polymeric
Bulk
500 m2/g
SiliaBond™
Silica Gel
SPE, For organic compounds from water, Drugs and metabolites from fluids
17%
60 Å
Column Chromatography
C18 (Octadecyl, ODS)
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Yes
Irregular
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit