Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaBond™ C8 (11% C) Monomeric, 40 to 63 μm, 60 Å
SiliaBond™ C8 (11% C) Monomeric, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.
Supplier: SiliCycle™ R30830B10G
Description
- Less retention compared to C18.
- For highly hydrophobic pesticides, small peptides and large molecule drugs.
Specifications
40 to 63 μm | |
C8 Monomeric | |
10 g | |
Keep dry. | |
Bulk Functionalized Irregular Silica Gel | |
Glass Gravity Columns | |
11% |
60 Å | |
Column Chromatography | |
C18 (Octadecyl, ODS) | |
SiliaBond™ | |
Silica Gel | |
Purification of large and highly hydrophobic molecules |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title