Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ C8 (11% C) Monomeric, 40 to 63 μm, 60 Å

SiliaBond™ C8 (11% C) Monomeric, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.

Supplier:  SiliCycle™ R30830B250G

 View more versions of this product

Catalog No. 01-182-201


Explore available promotions
Add to Cart

Description

Description

  • Less retention compared to C18.
  • For highly hydrophobic pesticides, small peptides and large molecule drugs.
Specifications

Specifications

40 to 63 μm
C8 Monomeric
250 g
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
11%
60 Å
Column Chromatography
C18 (Octadecyl, ODS)
SiliaBond™
Silica Gel
Purification of large and highly hydrophobic molecules
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.