Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Carbamate (CAR), 40 to 63 μm, 60 Å

Our Carbamate is a powerful scavenger for the removal of alcohols, alkoxides, amines, anilines, hydrazines, organometallics, and thiols.

Supplier:  SiliCycle™ R50130B100G

 View more versions of this product

Catalog No. 01-183-976


Add to cart

Description

Description

SiliaBond™ Carbamate acts as an Organic Scavenger for Nucleophiles. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our carbamate offers a great variety of formats for all purifications scales from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Covalent Bonding for Alcohols, Alkoxides, Amines, Anilines, Hydrazines, Organometallics, Thiols and Thiolates
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

Bulk Functionalized Irregular Silica Gel
100 g
SiliaBond™
Irregular
HDPE Bottle
0.741 g/mL
Safety and Handling

Safety and Handling

Recommended Storage : Keep Cool (<8°c) and Dry

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit