Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Carbamate (CAR), 40 to 63 μm, 60 Å
Our Carbamate is a powerful scavenger for the removal of alcohols, alkoxides, amines, anilines, hydrazines, organometallics, and thiols.
$39.65 - $2930.00
Specifications
Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
---|---|
Physical Form | Irregular |
Packaging | HDPE Bottle |
Product Line | SiliaBond™ |
Density | 0.741 g/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-183-972
|
SiliCycle™
R50130B5G |
5 g |
Each for $39.65
|
|
01-183-973
|
SiliCycle™
R50130B10G |
10 g |
Each for $61.70
|
|
01-183-974
|
SiliCycle™
R50130B25G |
25 g |
Each for $126.00
|
|
01-183-975
|
SiliCycle™
R50130B50G |
50 g |
Each for $229.00
|
|
01-183-976
|
SiliCycle™
R50130B100G |
100 g |
Each for $366.50
|
|
01-183-977
|
SiliCycle™
R50130B250G |
250 g |
Each for $859.00
|
|
01-183-978
|
SiliCycle™
R50130B500G |
500 g |
Each for $1,745.00
|
|
01-183-979
|
SiliCycle™
R50130B1KG |
1 kg |
Each for $2,930.00
|
|
Description
SiliaBond™ Carbamate acts as an Organic Scavenger for Nucleophiles. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our carbamate offers a great variety of formats for all purifications scales from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Covalent Bonding for Alcohols, Alkoxides, Amines, Anilines, Hydrazines, Organometallics, Thiols and Thiolates
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
HDPE Bottle | |
0.741 g/mL |
Irregular | |
SiliaBond™ |
Safety and Handling
Recommended Storage : Keep Cool (<8°c) and Dry
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.