Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Carboxylic Acid (WCX), 40 to 63 μm, 60 Å

Our Carboxylic Acid is a multifunctional gel that acts as a weak cation exchanger and organic scavenger for trapping primary and secondary amines and anilines.

Supplier:  SiliCycle™ R70030B250G

 View more versions of this product

Catalog No. 01-182-361


Add to cart

Description

Description

SiliaBond™ Carboxylic Acid acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. It offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, and is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment.

  • Weak cation exchanger of pKa of 4.8
  • Neutralized at pH below 2.8
  • For strong cationic species that may bind too strongly on SCX phases
  • Ionic Bonding and Catch and Release for Primary and Secondary Amines and Anilines
  • Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Carboxylic Acid (WCX)
Irregular Silica Gel
Carboxylic Acid (WCX)
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
Yes
Carboxylic Acid
60 Å
Column Chromatography
250 g
500 m2/g
SiliaBond™
Silica Gel
Bulk Scavenger in Post-reaction Mixture or Purification of Strong Basic Compounds
0.687 g/mL
HDPE Bottle
Irregular
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit