Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Carboxylic Acid (WCX), 40 to 63 μm, 60 Å
Our Carboxylic Acid is a multifunctional gel that acts as a weak cation exchanger and organic scavenger for trapping primary and secondary amines and anilines.
Supplier: SiliCycle™ R70030B50G
Description
SiliaBond™ Carboxylic Acid acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. It offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, and is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment.
- Weak cation exchanger of pKa of 4.8
- Neutralized at pH below 2.8
- For strong cationic species that may bind too strongly on SCX phases
- Ionic Bonding and Catch and Release for Primary and Secondary Amines and Anilines
- Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
40 to 63 μm | |
Carboxylic Acid (WCX) | |
Irregular Silica Gel | |
Carboxylic Acid (WCX) | |
Keep Dry | |
Bulk Functionalized Irregular Silica Gel | |
Bulk or Glass Gravity Columns | |
Irregular | |
Yes | |
Carboxylic Acid |
60 Å | |
Column Chromatography | |
50 g | |
500 m2/g | |
SiliaBond™ | |
Silica Gel | |
Bulk Scavenger in Post-reaction Mixture or Purification of Strong Basic Compounds | |
0.687 g/mL | |
HDPE Bottle | |
Irregular |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.