Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Diol nec, 40 to 63 μm, 60 Å

Our Diol is a multifunctional gel, a chromatographic phase and an organic scavenger for trapping boronic acids.

Supplier:  SiliCycle™ R35030B10G

 View more versions of this product

Catalog No. 01-182-247


Add to cart

Description

Description

SiliaBond™ Diol acts as a Chromatographic Phase and an Organic Scavenger for Electrophiles and Lewis Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our Diol offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • For difficult separation of low to medium polarity samples
  • Can be used in HILIC mode
  • For mono and polysaccharides separation
  • Covalent and Ionic Bonding for Boronic Acids
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Diol nec
Irregular Silica Gel
Diol (Dihydroxypropyl) nec
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
No
Irregular
60 Å
Column Chromatography
10 g
500 m2/g
SiliaBond™
Silica Gel
Purification of slightly polar compounds and HILIC mode
0.687 g/mL
HDPE Bottle
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit