Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel DMAP, 40 to 63 μm, 60 Å

Our DMAP is a heterogeneous reagent and organic scavenger for trapping acyl chlorides and sulfonyl chlorides.

Supplier:  SiliCycle™ R75630B25G

 View more versions of this product

Catalog No. 01-184-411


Add to cart

Description

Description

SiliaBond™ DMAP acts an Organic Scavenger for Electrophiles (Covalent Bonding for Acyl Chlorides and Sulfonyl Chlorides). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity.

  • Great variety of formats for all purifications scales, from laboratory to process to plant scales
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

HDPE Bottle
500 m2/g
25 g
Yes
Bulk or Glass Gravity Columns
DMAP
SiliaBond™
Irregular
40 to 63 μm
0.674 g/mL
Bulk Functionalized Irregular Silica Gel
Post-reaction Mixture or in Flash Chromatography
Irregular
60 Å
DMAP
Safety and Handling

Safety and Handling

Recommended Storage : Keep Cool (<8°C), Dry and Under Inert Atmosphere

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit