Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Guanidine (GUA), 40 to 63 μm, 60 Å
Our Guanidine is a powerful scavenger for the removal of acids, acidic phenols and boronic acids.
$104.10 - $5700.00
Specifications
Physical Form | Irregular |
---|---|
For Use With (Application) | Post-reaction Mixture or in Flash Chromatography |
Packaging | HDPE Bottle |
Particle Size | 40 to 63 μm |
Specific Surface Area | 500 m2/g |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-421
|
SiliCycle™
R68230B10G |
10 g |
Each for $104.10
|
|
|||||
01-184-432
|
SiliCycle™
R68230B25G |
25 g |
Each for $217.50
|
|
|||||
01-184-400
|
SiliCycle™
R68230B50G |
50 g |
Each for $398.00
|
|
|||||
01-184-419
|
SiliCycle™
R68230B100G |
100 g |
Each for $663.00
|
|
|||||
01-184-401
|
SiliCycle™
R68230B250G |
250 g |
Each for $1,513.00
|
|
|||||
01-184-402
|
SiliCycle™
R68230B500G |
500 g |
Each for $2,806.00
|
|
|||||
01-184-403
|
SiliCycle™
R68230B1KG |
1 kg |
Each for $5,700.00
|
|
|||||
01-184-399
|
SiliCycle™
R68230B5G |
5 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
SiliaBond™ Guanidine acts as an Organic Scavenger for Acids (Ionic Bonding and Catch and Release for Acids, Acidic Phenols and Boronic Acids). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity.
- Great variety of formats for all purifications scales, from laboratory to process to plant scales
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards, no safety precautions needed
Specifications
Irregular | |
HDPE Bottle | |
500 m2/g | |
Yes | |
Irregular | |
SiliaBond™ |
Post-reaction Mixture or in Flash Chromatography | |
40 to 63 μm | |
Bulk Functionalized Irregular Silica Gel | |
Bulk or Glass Gravity Columns | |
60 Å | |
Guanidine (GUA) |
Safety and Handling
Recommended Storage : Keep Dry
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title