Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2) nec, 40 to 63 μm, 60 Å

Propylsulfonic Acid (SCX-2) nec is a multifunctional gel which acts as a strong cation exchanger for catch and release of weak cations and removes Amines and Anilines.

Supplier:  SiliCycle™ R51430B100G

 View more versions of this product

Catalog No. 01-182-278


Add to cart

Description

Description

SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. This gel provides a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and has great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong cation exchangers of pKa <1, permanently negatively charged and pH independent; for catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
  • Ionic Bonding and Catch and Release for Amines and Anilines
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Propylsulfonic Acid (SCX-2) nec
Irregular Silica Gel
Propylsulfonic Acid (SCX-2) nec
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
No
Irregular
60 Å
Column Chromatography
100 g
500 m2/g
SiliaBond™
Silica Gel
Post-reaction Mixture or Extraction of Weak Basic Molecules, Catch and Release of Amines
0.728 g/mL
HDPE Bottle
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit