Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2) nec, 40 to 63 μm, 60 Å
Propylsulfonic Acid (SCX-2) nec is a multifunctional gel which acts as a strong cation exchanger for catch and release of weak cations and removes Amines and Anilines.
Supplier: SiliCycle™ R51430B500G
Description
SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. This gel provides a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and has great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong cation exchangers of pKa <1, permanently negatively charged and pH independent; for catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
- Ionic Bonding and Catch and Release for Amines and Anilines
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
40 to 63 μm | |
Propylsulfonic Acid (SCX-2) nec | |
Irregular Silica Gel | |
Propylsulfonic Acid (SCX-2) nec | |
Keep Dry | |
Bulk Functionalized Irregular Silica Gel | |
Bulk or Glass Gravity Columns | |
Irregular | |
No | |
Irregular |
60 Å | |
Column Chromatography | |
500 g | |
500 m2/g | |
SiliaBond™ | |
Silica Gel | |
Post-reaction Mixture or Extraction of Weak Basic Molecules, Catch and Release of Amines | |
0.728 g/mL | |
HDPE Bottle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.