Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel TMA Acetate (SAX-2) nec, 40 to 63 μm, 60 Å

Our TMA Acetate gel is a multifunctional chromatographic phase that acts as a strong anion exchanger with easily exchangeable acetate counter-ion.

Supplier:  SiliCycle™ R66430B250G

 View more versions of this product

Catalog No. 01-182-337


Add to cart

Description

Description

SiliaBond™ TMA Acetate (SAX-2) acts as a Chromatographic Phase and an Organic Scavenger for Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel has a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong anion exchangers with easily exchangeable acetate counter-ion - more than chloride ion - for compounds with pKa <5 such as carboxylic acids
  • Ionic Bonding and Catch and Release for Acids and Acidic Phenols
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with a sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
TMA Acetate (SAX-2) nec
Irregular Silica Gel
TMA Acetate (Tributylammonium Chloride, SAX-2) nec
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
No
Irregular
60 Å
Column Chromatography
250 g
500 m2/g
SiliaBond™
Silica Gel
Bulk scavenger in post-reaction mixture or purification of weak acidic molecules
0.665 g/mL
HDPE Bottle
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit