Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel TMA Acetate (SAX-2) nec, 40 to 63 μm, 60 Å
Our TMA Acetate gel is a multifunctional chromatographic phase that acts as a strong anion exchanger with easily exchangeable acetate counter-ion.
Supplier: SiliCycle™ R66430B500G
Description
SiliaBond™ TMA Acetate (SAX-2) acts as a Chromatographic Phase and an Organic Scavenger for Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel has a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong anion exchangers with easily exchangeable acetate counter-ion - more than chloride ion - for compounds with pKa <5 such as carboxylic acids
- Ionic Bonding and Catch and Release for Acids and Acidic Phenols
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with a sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
40 to 63 μm | |
TMA Acetate (SAX-2) nec | |
Irregular Silica Gel | |
TMA Acetate (Tributylammonium Chloride, SAX-2) nec | |
Keep Dry | |
Bulk Functionalized Irregular Silica Gel | |
Bulk or Glass Gravity Columns | |
Irregular | |
No | |
Irregular |
60 Å | |
Column Chromatography | |
500 g | |
500 m2/g | |
SiliaBond™ | |
Silica Gel | |
Bulk scavenger in post-reaction mixture or purification of weak acidic molecules | |
0.665 g/mL | |
HDPE Bottle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.