Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Tosyl Chloride (TsCl), 40 to 63 μm, 60 Å
Our Tosyl Chloride is a powerful scavenger for the removal of alcohols, alkoxides, amines, anilines, hydrazines, organometallics, and thiols.
$2350.00 - $5030.00
Specifications
Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
---|---|
Physical Form | Irregular |
Packaging | HDPE Bottle |
Product Line | SiliaBond™ |
Density | 0.761 g/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-183-953
|
SiliCycle™
R44030B250G |
250 g |
Each for $2,350.00
|
|
|||||
01-183-954
|
SiliCycle™
R44030B500G |
500 g |
Each for $3,070.00
|
|
|||||
01-183-955
|
SiliCycle™
R44030B1KG |
1 kg |
Each for $5,030.00
|
|
|||||
01-183-948
|
SiliCycle™
R44030B5G |
5 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
01-183-949
|
SiliCycle™
R44030B10G |
10 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
01-183-950
|
SiliCycle™
R44030B25G |
25 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
01-183-951
|
SiliCycle™
R44030B50G |
50 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
01-183-952
|
SiliCycle™
R44030B100G |
100 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
SiliaBond™ Tosyl Chloride acts as an Organic Scavenger for Nucleophiles (Covalent Bonding for Alcohols, Alkoxides, Amines, Anilines, Hydrazines, Organometallics, Thiols and Thiolates). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our tosyl chloride offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Successful technology for a variety of fields: pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
HDPE Bottle | |
0.761 g/mL |
Irregular | |
SiliaBond™ |
Safety and Handling
Recommended Storage : Keep Cool (<8°C), Dry and Under Inert Atmosphere
Your input is important to us. Please complete this form to provide feedback related to the content on this product.