Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gels, Glycidoxy (GLY) nec, 40 to 63 μm, 60 Å
Our Glycidoxy (Si-GLY) may be used as a linker for further modification of the surface.
Supplier:  SiliCycle™ R36030B250G
 View more versions of this product
Catalog No. 01-184-373
Add to cart
Description
Description
SiliaBond™ Glycidoxy (Si-GLY) may be used as a linker for further modification of the surface and for the immobilization of molecules bearing amino, hydroxy, mercapto and thiocarboxylic acid groups.
Specifications
Specifications
40 to 63 μm
Bulk Functionalized Irregular Silica Gel
Irregular
SiliaBond™
250 g
Surface Modification and Immobilization of Molecules with Specific Groups
60 Å
Glycidoxy (GLY) nec