Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gels, Pyridinium Dichromate (PDC), 40 to 63 μm, 60 Å
Our Pyridinium Dichromate (Si-PDC) may be used as an alternative to Si-PCC in nucleoside and carbohydrate oxidation.
Supplier: SiliCycle™ R24530B10G
Description
SiliaBond™ Pyridinium Dichromate (Si-PDC) may be used as an alternative to Si-PCC in nucleoside and carbohydrate oxidation, particularly for fragile molecules. SiliaBond™ PDC can also be used in conjunction with tertbutylhydroperoxide for a variety of oxidative transformations.
PDC immobilized onto silica is a very convenient and effective reagent for oxidizing allylic and benzylic alcohols, saturated primary and secondary alcohols, and is compatible with acid-sensitive groups, such as cyclopropane ring or ketal function.
Oxidation of 4-tert-butylcyclohexanol to 4-tert-butylcyclohexanone with 2 eq. of Si-PDC in anhydrous DCM at room temperature is complete after 6 hours.
Specifications
40 to 63 μm | |
Bulk Functionalized Irregular Silica Gel | |
Irregular | |
SiliaBond™ |
10 g | |
Oxidizing Allylic and Benzylic Alcohols, Saturated Primary and Secondary Alcohols | |
60 Å | |
Pyridinium Dichromate (PDC) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.