Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gels, Pyridinium Dichromate (PDC), 40 to 63 μm, 60 Å
Our Pyridinium Dichromate (Si-PDC) may be used as an alternative to Si-PCC in nucleoside and carbohydrate oxidation.
Supplier:  SiliCycle™ R24530B250G
 View more versions of this product
Catalog No. 01-184-366
Add to cart
Description
Description
SiliaBond™ Pyridinium Dichromate (Si-PDC) may be used as an alternative to Si-PCC in nucleoside and carbohydrate oxidation, particularly for fragile molecules. SiliaBond™ PDC can also be used in conjunction with tertbutylhydroperoxide for a variety of oxidative transformations.
PDC immobilized onto silica is a very convenient and effective reagent for oxidizing allylic and benzylic alcohols, saturated primary and secondary alcohols, and is compatible with acid-sensitive groups, such as cyclopropane ring or ketal function.
Oxidation of 4-tert-butylcyclohexanol to 4-tert-butylcyclohexanone with 2 eq. of Si-PDC in anhydrous DCM at room temperature is complete after 6 hours.
Specifications
Specifications
40 to 63 μm
Bulk Functionalized Irregular Silica Gel
Irregular
SiliaBond™
250 g
Oxidizing Allylic and Benzylic Alcohols, Saturated Primary and Secondary Alcohols
60 Å
Pyridinium Dichromate (PDC)