Learn More
SiliCycle™ SiliaBond™ Organic Nucleophile Complete Scavenger Kit: Carbonate, Carbamate, Tosyl Chloride
Because all matrices are unique and small differences can influence scavenging efficiency, our Organic Nucleophile Complete Scavenger Kit is the perfect starting point for screening your Scavenger.
Supplier: SiliCycle™ K32630B5G
Description
Organic Nucleophile Complete Scavenger Kit offers successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and has great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
The Kit is composed of the following Scavengers:
- CARBONATE (CO3) (Ionic Bonding/Catch and Release for Acids, Acidic Phenols and Boronic Acids)
- CARBAMATE (Covalent Bonding for Alcohols, Alkoxides, Amines, Anilines, Hydrazines, Organometallics, Thiols and Thiolates)
- MALEIMIDE (Covalent Bonding for Thiols and Thiolates)
- TOSYL CHLORIDE (Covalent Bonding for Alcohols, Alkoxides, Amines, Anilines, Hydrazines, Organometallics, Thiols and Thiolates)
- Only need to filter out the scavenger once it has trapped the residual metal or organic impurity
- Great variety of formats for all purifications scales, from laboratory to process to plant scales
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Powder | |
5 g | |
SiliaBond™ |
Irregular | |
HDPE Bottle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.