Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaBond™ Organic Reversed-Phase Kit
For your convenience, we have combined our most popular bulk reagents into the SiliaBond™ Organic Reversed-Phase Kit, so you can be prepared with material in hand for any application.
Supplier: SiliCycle™ K32830B10G
Description
- Our Kits are available in 5 g, 10 g, 25 g, 50 g or 100 g format.
- Includes: C8, C18, C18 nec, C18 WPD, Cyano and Phenyl
- All in 40 to 63 μm 60 Å
Specifications
40 to 63 μm | |
C8, C18, C18 nec, C18 WPD, Cyano, Phenyl | |
Bulk | |
Reversed-Phase Kit (6 different phases) | |
SiliaBond™ | |
Silica Gel | |
Screening of reversed-phases |
60 Å | |
Column Chromatography | |
10 g | |
Keep dry. | |
Bulk Functionalized Irregular Silica Gel | |
Glass Gravity Columns | |
Irregular |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title