Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaBond™ Pentafluorophenyl (PFP), 40 to 63 μm, 60 Å
SiliaBond™ Pentafluorophenyl (PFP), 40 - 63 μm, 60 Å is a phase with a new selectivity approach with aromatic ring interactions for the purification of conjugated compounds (isomers).
Supplier: SiliCycle™ R67530B10G
Description
- Sorbent primarily used in the separation of molecules bearing fluorine atoms but may also be used in the separation of non-fluorous compounds such as Taxol™ and its derivatives.
- Si-PFP has a higher selectivity for aromatics containing molecules compared to the other fluorinated sorbents due to its phenyl ring.
Specifications
40 to 63 μm | |
PFP (Pentafluorophenyl) | |
10 g | |
500 m2/g | |
SiliaBond™ | |
Silica Gel | |
Purification of polar and aromatic compounds, Separation of isomers | |
11% |
60 Å | |
Column Chromatography | |
PFP (Pentafluorophenyl) | |
Keep dry. | |
Bulk Functionalized Irregular Silica Gel | |
Glass Gravity Columns | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title