Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Pentafluorophenyl (PFP), 40 to 63 μm, 60 Å

SiliaBond™ Pentafluorophenyl (PFP), 40 - 63 μm, 60 Å is a phase with a new selectivity approach with aromatic ring interactions for the purification of conjugated compounds (isomers).

Supplier:  SiliCycle™ R67530B50G

 View more versions of this product

Catalog No. 01-182-356


Add to Cart

Description

Description

  • Sorbent primarily used in the separation of molecules bearing fluorine atoms but may also be used in the separation of non-fluorous compounds such as Taxol™ and its derivatives.
  • Si-PFP has a higher selectivity for aromatics containing molecules compared to the other fluorinated sorbents due to its phenyl ring.
Specifications

Specifications

40 to 63 μm
PFP (Pentafluorophenyl)
50 g
500 m2/g
SiliaBond™
Silica Gel
Purification of polar and aromatic compounds, Separation of isomers
11%
60 Å
Column Chromatography
PFP (Pentafluorophenyl)
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Yes
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit