Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Phenyl (PHE), 40 to 63 μm, 60 Å

SiliaBond™ Phenyl (PHE), 40 - 63 μm, 60 Å is a moderate non-polar sorbent with different selectivity for aromatic compounds in comparison to other non-polar sorbents.

Supplier:  SiliCycle™ R34030B500G

 View more versions of this product

Catalog No. 01-182-243


Add to Cart

Description

Description

  • Moderately non-polar sorbent.
  • Alternative selectivity for aromatic compounds, compared to other reversed-phases.
Specifications

Specifications

40 to 63 μm
Phenyl
Bulk
Phenyl (C6H5)
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Yes
60 Å
Column Chromatography
500 g
500 m2/g
SiliaBond™
Silica Gel
Purification of aromatic and non-polar compounds
9%
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit