Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Phenyl (PHE), 40 to 63 μm, 60 Å

SiliaBond™ Phenyl (PHE), 40 - 63 μm, 60 Å is a moderate non-polar sorbent with different selectivity for aromatic compounds in comparison to other non-polar sorbents.

$33.10 - $2480.00

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Phenyl
Technique Column Chromatography
Packaging Bulk
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-182-245
View Documents
SiliCycle™
R34030B5G
5 g
Each for $33.10
Add to Cart
 
01-182-239
View Documents
SiliCycle™
R34030B10G
10 g
Each for $52.30
Add to Cart
 
01-182-242
View Documents
SiliCycle™
R34030B25G
25 g
Each for $105.00
Add to Cart
 
01-182-244
View Documents
SiliCycle™
R34030B50G
50 g
Each for $195.30
Add to Cart
 
01-182-238
View Documents
SiliCycle™
R34030B100G
100 g
Each for $312.00
Add to Cart
 
01-182-241
View Documents
SiliCycle™
R34030B250G
250 g
Each for $729.00
Add to Cart
 
01-182-243
View Documents
SiliCycle™
R34030B500G
500 g
Each for $1,376.00
Add to Cart
 
01-182-240
View Documents
SiliCycle™
R34030B1KG
1 kg
Each for $2,480.00
Add to Cart
 
Description

Description

  • Moderately non-polar sorbent.
  • Alternative selectivity for aromatic compounds, compared to other reversed-phases.
Specifications

Specifications

40 to 63 μm
Phenyl
Bulk
500 m2/g
SiliaBond™
Silica Gel
Purification of aromatic and non-polar compounds
9%
60 Å
Column Chromatography
Phenyl (C6H5)
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Yes
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit