Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Silver Nitrate, 60-200 μm, 60 Å

SiliaBond™ Silver Nitrate, 60-200 μm, 60 Å is mainly used for the separation of cis/trans isomers of unsaturated compounds such as alkenes, lipids, steroids and terpenes.

Supplier:  SiliCycle™ R23540B5G

 View more versions of this product

Catalog No. 01-182-196


Add to cart

Description

Description

  • For separation of cis/trans isomers of unsaturated compounds (alkenes, lipids, steroids and terpenes)
Specifications

Specifications

60 to 200 μm
Silver Nitrate (AgNO3)
Bulk
5 g
Light sensitive, store away from light.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
No
60 Å
Column Chromatography
2 to 10
Silver Nitrate (AgNO3, 10%)
SiliaBond™
Silica Gel
For the separation of cis/trans isomers of unsaturated compounds such as alkenes, lipids, steroids and terpenes
Irregular
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit