Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Tridecafluoro (TDF), 40 to 63 μm, 60 Å

SiliaBond™ Tridecafluoro (TDF), 40 - 63 μm, 60 Å is a chromatographic phase for fast and easy separation of fluorine containing molecules from non-fluorous ones.

Supplier:  SiliCycle™ R63530B500G

 View more versions of this product

Catalog No. 01-182-323


Add to Cart

Description

Description

  • Sorbent primarily used to separate fluorinated compounds.
  • It can also be used in fluorous solid-phase extraction (FSPE) or fluorous-tag compounds as reported for the synthesis of oligosaccharide.
Specifications

Specifications

40 to 63 μm
Tridecafluoro (TDF)
500 g
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Yes
60 Å
Column Chromatography
Tridecafluoro (TDF)
SiliaBond™
Silica Gel
Separation of fluorine containing molecules from non-fluorous ones
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit