Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaBond™ Triethylamine (WAX-2), 40 to 63 μm, 60 Å

SiliaBond™ Triethylamine (WAX-2), 40 - 63 μm, 60 Å is an excellent sorbent for extraction of strong acidic molecules and for catch and release of compounds bearing a permanant negative charge.

$41.25 - $3100.00

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Triethylamine (TEA, WAX-2)
Technique Column Chromatography
Packaging Bulk
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-182-373
View Documents
SiliCycle™
R76530B5G
5 g
Each for $41.25
Add to Cart
 
01-182-367
View Documents
SiliCycle™
R76530B10G
10 g
Each for $65.90
Add to Cart
 
01-182-370
View Documents
SiliCycle™
R76530B25G
25 g
Each for $129.20
Add to Cart
 
01-182-372
View Documents
SiliCycle™
R76530B50G
50 g
Each for $243.00
Add to Cart
 
01-182-366
View Documents Promotion Details
SiliCycle™
R76530B100G
100 g
Each for $388.50
Add to Cart
 
01-182-369
View Documents
SiliCycle™
R76530B250G
250 g
Each for $911.50
Add to Cart
 
01-182-371
View Documents
SiliCycle™
R76530B500G
500 g
Each for $1,719.00
Add to Cart
 
01-182-368
View Documents
SiliCycle™
R76530B1KG
1 kg
Each for $3,100.00
Add to Cart
 
Description

Description

  • Weak anion exchanger (pKa of 10.5), positively charged at pH below 8.5.
  • For catch and release of compounds bearing a permanent negative charge (like salts of sulfonic acids).
Specifications

Specifications

40 to 63 μm
Triethylamine (TEA, WAX-2)
Bulk
Triethylamine (TEA, WAX-2)
SiliaBond™
Silica Gel
Purification of strong acidic molecules
Bulk
60 Å
Column Chromatography
2 to 10
Keep dry and cool (≤ 8°C).
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Yes
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit