Learn More
SiliCycle™ SiliaChrom Plus Holders for Guard Cartridges
Holders for Guard Cartridges protect both analytical and preparative HPLC columns and are easy to use.
Supplier:  SiliCycle™ HPHQ010
 View more versions of this product
Catalog No. 01-184-219
Add to Cart
Description
Description
SiliaChrom Plus HPLC Guard Cartridges are designed to effectively protect both analytical and preparative HPLC columns. The use of these cartridges is highly recommended to protect and prolong column lifetime. SiliaChrom Plus Guard Cartridges are extremely cost effective and easy to use as a pre-filter to remove contaminants prior to your injections. Contaminants have an extremely high impact and can cause the following:
  • Higher back-pressure
  • Loss of resolution
  • Peak shape alterations (tailing or peak splitting)
  • Irreversible damages to column and/or system
  • Baseline noise or drift
To save you time and money, SiliaChrom Plus HPLC Guard Cartridges help prevent these issues in addition to having a limited influence on your chromatography.
Holders for Guard Cartridges are easy to use- simply insert the cartridge inside the metallic holder and tighten both parts of the holder together.
Specifications
Specifications
1
HPLC Guard Cartridge Holder
10 mm ID HPLC Columns
Semi-preparative Guard Holder
HPLC
SiliaChrom
10 mm
10 mm
Stainless Steel
Removal of Contaminants Prior to HPLC Injections
Guard Holder