Learn More
SiliCycle™ SiliaChrom Plus HPLC Guard Cartridges, Palladium Scavenger (DMT and Thiol), 5 μm
Crude reaction mixtures can directly be injected without further metal removal: these Palladium Scavenging Guard Cartridges will protect your HPLC column and your system from metallic impurities.
Supplier:  SiliCycle™ HPLGK34605EAQ010
 View more versions of this product
Catalog No. 01-184-226
Add to cart
Description
Description
SiliaChrom Plus HPLC Guard Cartridges are designed to effectively protect both analytical and preparative HPLC columns. The use of these cartridges is highly recommended to protect and prolong column lifetime. SiliaChrom Plus Guard Cartridges are extremely cost effective and easy to use as a pre-filter to remove contaminants prior to your injections. Contaminants have an extremely high impact and can cause the following:
  • Higher back-pressure
  • Loss of resolution
  • Peak shape alterations (tailing or peak splitting)
  • Irreversible damages to column and/or system
  • Baseline noise or drift
To save you time and money, SiliaChrom Plus HPLC Guard Cartridges help prevent these issues in addition to having a limited influence on your chromatography.
Specifications
Specifications
2 Pk.
5 μm
100 Å
10 mm
DMT and Thiol
Spherical, Fully Porous, Ultrapure Silica
Semi-preparative Guard Cartridge
HPLC, Metal Scavenging
Spherical
SiliaChrom
2 to 8
10 mm
HPLC Guard Cartridge
10 mm ID HPLC Columns
Silica Gel
Metal Scavenging
Removal of Metallic Contaminants Prior to HPLC Injections
Metal Scavenging
Yes