Learn More
SiliCycle™ SiliaFlash™ Irregular Silica Gel, 120 to 200 μm, 60 Å
Particles for Gravity Chromatography: silica for standard open column chromatography. Narrow particle size distribution enables uniform packing, Suitable for mass overload purification. Suitable for Peptides (1000 < MW < 10,000 Dalton)
Supplier: SiliCycle™ R10160B10KG
Description
SiliaFlash™ - Irregular Silica Gels for Chromatography
With pore diameters ranging from 30 to 300 Ångström (Å) and particle sizes up to 1200 μm, SiliCycle™ offers products to meet all your requirements.
SiliCycle™ offers one of the most reliable portfolios for flash and gravity grades for low to medium-high pressure. Their silica gels are ideal for preparative chromatography, from laboratory to pilot-plant processes and production scale. In addition to performance, SiliCycle™’s silica gels also ensure consistency, reliability and reproducibility.
Ensure Unbeatable Chromatography Performance:
- High purity silica with the lowest level of fines on the market
- No contamination, lower back-pressure and superior separation
- Exceptional narrow particle and pore size distributions
- Optimal separation and resolution
- Batch-to-batch, year-to-year consistency
- Reliable chromatography
- Neutral pH
- Wide range of products can be purified, even acid sensitive ones
- Low metal content & controlled water content
- Symmetrical peaks without tailing
- High mechanical stability
- Can be used under high pressures without surface abrasion
- High surface area and density
- Greater loading capacity, enabling more silica for the same volume
- Solvent economy (smaller dead volume)
- Availability in bulk quantities
- Always in stock with on-time delivery
Different grades available for SiliCycle™'s most popular products:
Over the years, SiliCycle™ has developed two different grades ('Standard' and 'Superior') for the two most popular irregular gels used in the industry: 40 to 63 μm, 60 Å and 60 to 200 μm, 60 Å
Those two grades of each gel are available to address all of SiliCycle™'s customers’ requirements, depending on their applications, areas of research, budgets and so on.
1. 40 to 63 μm, 60 Å silica gels
- Superior Grade (F60 PN: R10030B)
- Extra step to reduce metal content to minimum level
- Tighter particle size distribution
- Fines have been removed
- Standard Grade (P60 PN: R12030B)
- Fines have been removed
- Lower price
Both compare favorably with the overall industry average of a 40 to 63 μm distribution, and each grade offers its own particle size distribution profile.
2. 60 to 200 μm, 60 Å silica gels
- Superior Grade (G60 PN: R10040B)
- Extra step to reduce metal content to minimum level
- Tighter particle size distribution
- Fines have been removed
- Standard Grade (GE60 PN: R10140B)
- Fines have been removed
Each grade offers its own particle size distribution profile.
Specifications
120 to 200 μm | |
Silica | |
Bulk | |
Bare Silica (Silicon Dioxyde, SiO2) | |
SiliaFlash™ | |
Silica Gel | |
7631-86-9 |
60 Å | |
Column Chromatography | |
10 kg | |
Keep dry | |
Bulk Irregular Silica Gel | |
Chromatography, purification | |
Irregular |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Keep dry