Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaFlash™ Irregular Silica Gel, 500 to 1000 μm, 60 Å

Particles for other applications, such as silica for plugs. Suitable for Organic Molecules (MW <1000 Dalton).

Supplier:  SiliCycle™ R10180B1KG

 View more versions of this product

Catalog No. 01-182-495


Add to cart

Description

Description

SiliaFlash - Irregular Silica Gels for Chromatography

With pore diameters ranging from 30 to 300 Ångström (Å) and particle sizes up to 1200 μm, SiliCycle™ offers products to meet all your requirements.

SiliCycle™ offers one of the most reliable portfolios for flash and gravity grades for low to medium-high pressure. Their silica gels are ideal for preparative chromatography, from laboratory to pilot-plant processes and production scale. In addition to performance, SiliCycle™’s silica gels also ensure consistency, reliability and reproducibility.

Ensure Unbeatable Chromatography Performance:

  • High purity silica with the lowest level of fines on the market
    • No contamination, lower back-pressure and superior separation
  • Exceptional narrow particle and pore size distributions
    • Optimal separation and resolution
  • Batch-to-batch, year-to-year consistency
    • Reliable chromatography
  • Neutral pH
    • Wide range of products can be purified, even acid sensitive ones
  • Low metal content & controlled water content
    • Symmetrical peaks without tailing
  • High mechanical stability
    • Can be used under high pressures without surface abrasion
  • High surface area and density
    • Greater loading capacity, enabling more silica for the same volume
    • Solvent economy (smaller dead volume)
  • Availability in bulk quantities
    • Always in stock with on-time delivery

Different grades available for SiliCycle™'s most popular products:

Over the years, SiliCycle™ has developed two different grades ('Standard' and 'Superior') for the two most popular irregular gels used in the industry: 40 to 63 μm, 60 Å and 60 to 200 μm, 60 Å

Those two grades of each gel are available to address all of SiliCycle™'s customers’ requirements, depending on their applications, areas of research, budgets and so on.

 1. 40 to 63 μm, 60 Å silica gels

  • Superior Grade (F60 PN: R10030B)
    • Extra step to reduce metal content to minimum level
    • Tighter particle size distribution
    • Fines have been removed
  • Standard Grade (P60 PN: R12030B)
    • Fines have been removed
    • Lower price

Both compare favorably with the overall industry average of a 40 to 63 μm distribution, and each grade offers its own particle size distribution profile.

2. 60 to 200 μm, 60 Å silica gels

  • Superior Grade (G60 PN: R10040B)
    • Extra step to reduce metal content to minimum level
    • Tighter particle size distribution
    • Fines have been removed
  • Standard Grade (GE60 PN: R10140B)
    • Fines have been removed

Each grade offers its own particle size distribution profile.

Specifications

Specifications

500 to 1000 μm
Silica
Bulk
Bare Silica (Silicon Dioxyde, SiO2)
SiliaFlash™
Silica Gel
7631-86-9
60 Å
Column Chromatography
1 kg
Keep dry
Bulk Irregular Silica Gel
Chromatography, purification
Irregular
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Keep dry