Learn More
SiliCycle™ SiliaMetS™ E-PAK™ C-CA Activated Carbon
Our C-CA is a chemically activated, wood-based powdered activated carbon.
Supplier:  SiliCycle™ CCA5KG
 View more versions of this product
Catalog No. 01-182-153
Add to cart
Description
Description
SiliaCarb CA is an activated carbon with high adsorptive capacity for high molecular weight organics. Its high surface area and a large volume of mesopores allow it to rapidly remove color, taste, and odor compounds from solution.
  • Chemically activated
  • Wood-based
Specifications
Specifications
Black
5 kg
E-PAK
5 to 9
Powder
Removal of precious metal catalysts, pigments and reaction contaminants