Learn More
SiliCycle™ SiliaMetS™ E-PAK™ C-CA Activated Carbon
Our C-CA is a chemically activated, wood-based powdered activated carbon.
Supplier: SiliCycle™ CCA20KG
Description
SiliaCarb CA is an activated carbon with high adsorptive capacity for high molecular weight organics. Its high surface area and a large volume of mesopores allow it to rapidly remove color, taste, and odor compounds from solution.
- Chemically activated
- Wood-based
Specifications
Black | |
20 kg | |
E-PAK |
5 to 9 | |
Powder | |
Removal of precious metal catalysts, pigments and reaction contaminants |