Learn More
SiliCycle™ SiliaMetS™ E-PAK™ C-HA Activated Carbon
Our C-HA is a high-activity, wood-based powdered activated carbon.
Supplier:  SiliCycle™ CHA20KG
 View more versions of this product
Catalog No. 01-182-163
Add to Cart
Description
Description
SiliaCarb HA is a powdered activated carbon with unusually high adsorptive capacity for a wide range of organic compounds. SiliaCarb HA is specially processed to produce a high-purity product with low ash content, low soluble iron, and a pore structure that is optimum for a wide range of applications.
  • High-activity
  • Wood-based
Specifications
Specifications
Black
20 kg
E-PAK
5 to 8
Powder
Removal of precious metal catalysts, pigments and reaction contaminants